Assay Details
Target Gene Details
Entrez Gene ID: | 598 |
Gene Name: | BCL2 like 1 |
Gene Aliases: |
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52 |
Location: |
Chr.20:31664452-31723999 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L1 | NM_001191.3 | 3 | 1376 | NP_001182.1 |
| NM_001317919.1 | 3 | 1547 | NP_001304848.1 | |
| NM_001317920.1 | 3 | 1202 | NP_001304849.1 | |
| NM_001317921.1 | 3 | 1331 | NP_001304850.1 | |
| NM_001322239.1 | 3 | 1220 | NP_001309168.1 | |
| NM_001322240.1 | 3 | 1349 | NP_001309169.1 | |
| NM_001322242.1 | 3 | 1202 | NP_001309171.1 | |
| NM_138578.2 | 3 | 1565 | NP_612815.1 | |
| NR_134257.1 | 3 | 1223 | ||
| XM_011528964.2 | 3 | 1184 | XP_011527266.1 | |
| XM_017027993.1 | 3 | 1190 | XP_016883482.1 | |
| BC019307.1 | 3 | 1320 | AAH19307.1 | |
| BX647525.1 | 5 | 1101 | ||
| CR936637.1 | 3 | 1351 | CAI56777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
| nsv585911 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map