Assay Details
Target Gene Details
Entrez Gene ID: | 9811 |
Gene Name: | cap binding complex dependent translation initiation factor |
Gene Aliases: |
Gm672, KIAA0427 |
Location: |
Chr.18:48539056-48863217 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTIF | NM_001142397.1 | NP_001135869.1 | ||
| NM_014772.2 | NP_055587.1 | |||
| XM_005258392.4 | XP_005258449.1 | |||
| XM_006722583.3 | XP_006722646.1 | |||
| XM_006722586.3 | XP_006722649.1 | |||
| XM_006722587.3 | XP_006722650.1 | |||
| XM_006722588.3 | XP_006722651.1 | |||
| XM_006722589.3 | XP_006722652.1 | |||
| XM_011526278.2 | XP_011524580.1 | |||
| XM_011526279.2 | XP_011524581.1 | |||
| XM_017026100.1 | XP_016881589.1 | |||
| XM_017026101.1 | XP_016881590.1 | |||
| XM_017026102.1 | XP_016881591.1 | |||
| AB007887.1 | ||||
| AK095970.1 | ||||
| BC042146.1 | AAH42146.1 |
Target Gene Details
Entrez Gene ID: | 100616366 |
Gene Name: | microRNA 4743 |
Gene Aliases: |
- |
Location: |
Chr.18:48670600-48670668 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4743 | NR_039897.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576857 | Chr.18:48658158 - 48674681 on Build GRCh38 | Loss |
|
| nsv509694 | Chr.18:48660149 - 48687052 on Build GRCh38 | Insertion |
|
| nsv2291 | Chr.18:48659613 - 48685659 on Build GRCh38 | Insertion |
|
| nsv833642 | Chr.18:48449936 - 48694680 on Build GRCh38 | Gain |
|
| nsv833643 | Chr.18:48615354 - 48771229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map