Assay Details
Target Gene Details
Entrez Gene ID: | 57680 |
Gene Name: | chromodomain helicase DNA binding protein 8 |
Gene Aliases: |
AUTS18, HELSNF1 |
Location: |
Chr.14:21385194-21437298 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD8 | NM_001170629.1 | 29 | 5396 | NP_001164100.1 |
| NM_020920.3 | 30 | 4641 | NP_065971.2 | |
| AB046784.3 | 29 | 4884 | BAB13390.3 | |
| AK056639.1 | 10 | 1263 | ||
| AK122910.1 | 30 | 4589 | ||
| AK131077.1 | 15 | 2220 | BAC85127.1 | |
| BC098452.1 | 4 | 312 | AAH98452.1 | |
| CR749315.1 | 30 | 4586 | CAH18170.1 |
More Information
Set Membership: |
|

Genomic Map