Assay Details
Target Gene Details
Entrez Gene ID: | 26277 |
Gene Name: | TERF1 interacting nuclear factor 2 |
Gene Aliases: |
DKCA3, TIN2 |
Location: |
Chr.14:24239641-24242674 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TINF2 | NM_001099274.1 | 9 | 1654 | NP_001092744.1 |
| NM_012461.2 | 6 | 1981 | NP_036593.2 | |
| XM_005267529.3 | 8 | 1480 | XP_005267586.1 | |
| XM_011536642.2 | 5 | 1876 | XP_011534944.1 | |
| XM_017021216.1 | 5 | 1186 | XP_016876705.1 | |
| XM_017021217.1 | 5 | 778 | XP_016876706.1 | |
| AF195512.1 | 6 | 1901 | AAF18439.1 | |
| AK023166.1 | 4 | 2372 | BAB14440.1 | |
| AK225379.1 | 6 | 1925 | ||
| AK294119.1 | 8 | 1480 | ||
| BC005030.1 | 9 | 1435 | AAH05030.1 | |
| BC019343.1 | 9 | 1612 | AAH19343.1 | |
| BX161478.1 | 4 | 1709 | CAD61933.1 | |
| EU851975.1 | 6 | 828 |
More Information
Set Membership: |
|

Genomic Map