Assay Details
Target Gene Details
Entrez Gene ID: | 7398 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
UBP |
Location: |
Chr.1:62436304-62451804 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP1 | NM_001017415.1 | 6 | 1522 | NP_001017415.1 |
| NM_001017416.1 | 6 | 1427 | NP_001017416.1 | |
| NM_003368.4 | 6 | 1952 | NP_003359.3 | |
| AB014458.1 | 6 | 1303 | BAA34703.1 | |
| AB208893.1 | 6 | 1584 | BAD92130.1 | |
| AF117386.1 | 6 | 1439 | AAD11441.1 | |
| AK312423.1 | 6 | 1615 | ||
| AL117575.1 | 5 | 1274 | CAB55999.1 | |
| BC050525.1 | 6 | 1656 | AAH50525.1 |
More Information
Set Membership: |
|

Genomic Map