Assay Details
Target Gene Details
Entrez Gene ID: | 64927 |
Gene Name: | tetratricopeptide repeat domain 23 |
Gene Aliases: |
HCC-8 |
Location: |
Chr.15:99136323-99251226 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055008 | Chr.15:98933915 - 99479656 on Build GRCh38 | Gain |
|
| nsv570716 | Chr.15:99086672 - 99376127 on Build GRCh38 | Gain |
|
| esv3892741 | Chr.15:98979139 - 99618239 on Build GRCh38 | Gain |
|
| nsv1045124 | Chr.15:98961614 - 99307005 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map