Assay Details
Target Gene Details
Entrez Gene ID: | 84864 |
Gene Name: | MYC induced nuclear antigen |
Gene Aliases: |
MDIG, MINA53, NO52, ROX |
Location: |
Chr.3:97941817-97972451 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MINA | NM_001042533.2 | 10 | 1900 | NP_001035998.1 |
| NM_001261829.1 | 10 | 1430 | NP_001248758.1 | |
| NM_032778.5 | 10 | 1897 | NP_116167.3 | |
| NM_153182.3 | 10 | 1433 | NP_694822.2 | |
| XM_005247838.4 | 10 | 1650 | XP_005247895.1 | |
| XM_011513244.2 | 6 | 1162 | XP_011511546.1 | |
| AB083189.1 | 10 | 1536 | BAC16537.1 | |
| AB083190.1 | 10 | 1533 | BAC16358.1 | |
| AK027299.1 | 10 | 1380 | BAB55024.1 | |
| AY302110.3 | 10 | 1603 | AAP59421.1 | |
| AY390536.1 | 10 | 1585 | AAR27293.1 | |
| AY456380.1 | 10 | 1167 | AAR21572.1 | |
| BC014928.1 | 10 | 1587 | AAH14928.1 | |
| CR627479.1 | 9 | 1186 | CAH10679.1 | |
| DQ453796.1 | 10 | 1397 | ABE28016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520553 | Chr.3:95905623 - 98010484 on Build GRCh38 | Gain |
|
| nsv1013060 | Chr.3:96856275 - 98493673 on Build GRCh38 | Gain |
|
| nsv1012322 | Chr.3:97894388 - 97985590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map