Assay Details
Target Gene Details
Entrez Gene ID: | 57410 |
Gene Name: | SCY1 like pseudokinase 1 |
Gene Aliases: |
GKLP, HT019, NKTL, NTKL, P105, SCAR21, TAPK, TEIF, TRAP |
Location: |
Chr.11:65525065-65538711 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCYL1 | NM_001048218.1 | NP_001041683.1 | ||
| NM_020680.3 | NP_065731.3 | |||
| XM_005274118.3 | XP_005274175.1 | |||
| XM_005274120.3 | XP_005274177.1 | |||
| XM_005274121.3 | XP_005274178.1 | |||
| XM_017018029.1 | XP_016873518.1 | |||
| XM_017018030.1 | XP_016873519.1 | |||
| XM_017018031.1 | XP_016873520.1 | |||
| XM_017018032.1 | XP_016873521.1 | |||
| XM_017018033.1 | XP_016873522.1 | |||
| AB047077.1 | BAB55454.1 | |||
| AB051427.1 | BAB55458.1 | |||
| AB051428.1 | BAB55459.1 | |||
| AF225424.1 | AAG09726.1 | |||
| AF297709.1 | AAG17902.1 | |||
| AK302198.1 | ||||
| AK314945.1 | ||||
| BC009967.2 | AAH09967.2 | |||
| BC069233.1 | AAH69233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv361 | Chr.11:65520835 - 65534840 on Build GRCh38 | Insertion |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map