Assay Details
Target Gene Details
Entrez Gene ID: | 6648 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD |
Location: |
Chr.6:159679064-159762529 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOD2 | NM_000636.3 | 5 | 815 | NP_000627.2 |
| NM_001024465.2 | NP_001019636.1 | |||
| NM_001024466.2 | NP_001019637.1 | |||
| NM_001322814.1 | 4 | 698 | NP_001309743.1 | |
| NM_001322815.1 | 4 | 635 | NP_001309744.1 | |
| NM_001322817.1 | NP_001309746.1 | |||
| NM_001322819.1 | 5 | 721 | NP_001309748.1 | |
| NM_001322820.1 | 5 | 724 | NP_001309749.1 | |
| AK097395.1 | ||||
| AK296809.1 | 4 | 611 | ||
| AK304766.1 | 4 | 548 | ||
| AK313082.1 | ||||
| AY280718.1 | AAP34407.1 | |||
| AY280719.1 | AAP34408.1 | |||
| AY280720.1 | AAP34409.1 | |||
| AY280721.1 | AAP34410.1 | |||
| BC012423.1 | AAH12423.1 | |||
| BM724413.1 | ||||
| BM994509.1 | ||||
| BT006967.1 | AAP35613.1 | |||
| BU741675.1 | ||||
| DQ003134.1 | AAY21807.1 | |||
| M36693.1 | 5 | 748 | AAA36622.1 | |
| X07834.1 | 5 | 658 | CAA30687.1 | |
| X14322.1 | 5 | 749 | CAA32502.1 | |
| X15132.1 | 5 | 714 | CAA33228.1 | |
| X59445.1 | 5 | 714 | CAA42066.1 | |
| Y00472.1 | CAA68533.1 | |||
| Y00985.1 | 5 | 748 | CAA68791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map