Assay Details
Target Gene Details
Entrez Gene ID: | 4504 |
Gene Name: | metallothionein 3 |
Gene Aliases: |
GIF, GIFB, GRIF, ZnMT3 |
Location: |
Chr.16:56589355-56591088 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MT3 | NM_005954.2 | 3 | 382 | NP_005945.1 |
| BC013081.1 | 3 | 193 | AAH13081.1 | |
| BT007030.1 | 3 | 146 | AAP35677.1 | |
| CB155600.1 | 3 | 240 | ||
| D13365.1 | 3 | 198 | ||
| X89604.1 | 3 | 146 | CAA61763.1 |
More Information
Set Membership: |
|

Genomic Map