Assay Details
Target Gene Details
Entrez Gene ID: | 101929516 |
Gene Name: | uncharacterized LOC101929516 |
Gene Aliases: |
- |
Location: |
Chr.1:39565070-39573203 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929516 | NR_125979.1 | |||
| DB086403.1 | ||||
| HY203033.1 |
Target Gene Details
Entrez Gene ID: | 8761 |
Gene Name: | poly(A) binding protein cytoplasmic 4 |
Gene Aliases: |
APP-1, APP1, PABP4, iPABP |
Location: |
Chr.1:39560813-39576849 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PABPC4 | NM_001135653.1 | NP_001129125.1 | ||
| NM_001135654.1 | NP_001129126.1 | |||
| NM_003819.3 | NP_003810.1 | |||
| BC065540.1 | AAH65540.1 | |||
| BC071591.1 | AAH71591.1 | |||
| BC094755.1 | AAH94755.1 | |||
| BC106908.1 | AAI06909.1 | |||
| BC118568.1 | ||||
| D12799.1 | BAA02244.1 | |||
| U33818.1 | AAC50350.1 | |||
| U75686.1 | AAB97309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829649 | Chr.1:39505283 - 39638842 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map