Assay Details
Target Gene Details
Entrez Gene ID: | 283807 |
Gene Name: | F-box and leucine rich repeat protein 22 |
Gene Aliases: |
Fbl22 |
Location: |
Chr.15:63596225-63608898 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL22 | NM_203373.2 | 1 | 100 | NP_976307.2 |
| XM_005254320.3 | 1 | 1228 | XP_005254377.1 | |
| XM_011521468.2 | 1 | 66 | XP_011519770.1 | |
| XM_011521469.1 | 1 | 62 | XP_011519771.1 | |
| XM_011521472.2 | 1 | 66 | XP_011519774.1 | |
| XM_017022086.1 | 1 | 1228 | XP_016877575.1 | |
| AA495872.1 | 1 | 100 | ||
| AK096945.1 | 1 | 62 | ||
| BC065833.1 | 1 | 56 | AAH65833.1 |
Target Gene Details
Entrez Gene ID: | 100130855 |
Gene Name: | USP3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:63587710-63600827 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3-AS1 | NR_034080.1 | |||
| AK093077.1 | ||||
| DB444752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map