Assay Details
Target Gene Details
Entrez Gene ID: | 56147 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
PCDH-ALPHA1 |
Location: |
Chr.5:140786136-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA1 | NM_018900.3 | NP_061723.1 | ||
| NM_031411.2 | NP_113599.1 | |||
| AF152305.1 | AAD43699.1 | |||
| AF169695.1 | AAF89692.1 |
Target Gene Details
Entrez Gene ID: | 56139 |
Gene Name: | protocadherin alpha 10 |
Gene Aliases: |
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10 |
Location: |
Chr.5:140855883-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA10 | NM_018901.3 | NP_061724.1 | ||
| NM_031860.2 | NP_114066.1 | |||
| AF152306.1 | AAD43700.1 | |||
| AF169689.1 | AAF89329.1 | |||
| BC130447.1 |
Target Gene Details
Entrez Gene ID: | 56138 |
Gene Name: | protocadherin alpha 11 |
Gene Aliases: |
CNR7, CNRN7, CNRS7, CRNR7, PCDH-ALPHA11 |
Location: |
Chr.5:140867513-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA11 | NM_018902.4 | NP_061725.1 | ||
| AF152307.1 | AAD43701.1 | |||
| AJ007609.1 | CAC22256.1 |
Target Gene Details
Entrez Gene ID: | 56137 |
Gene Name: | protocadherin alpha 12 |
Gene Aliases: |
PCDH-ALPHA12 |
Location: |
Chr.5:140875302-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA12 | NM_018903.3 | NP_061726.1 | ||
| AF152308.1 | AAD43702.1 | |||
| AK075556.1 |
Target Gene Details
Entrez Gene ID: | 56136 |
Gene Name: | protocadherin alpha 13 |
Gene Aliases: |
CNR5, CNRN5, CNRS5, CRNR5, PCDH-ALPHA13 |
Location: |
Chr.5:140882269-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA13 | NM_018904.2 | NP_061727.1 | ||
| AB208924.1 | BAD92161.1 | |||
| AF152309.1 | AAD43703.1 |
Target Gene Details
Entrez Gene ID: | 56146 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
PCDH-ALPHA2 |
Location: |
Chr.5:140794859-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA2 | NM_018905.2 | NP_061728.1 | ||
| AF152310.1 | AAD43704.1 |
Target Gene Details
Entrez Gene ID: | 56145 |
Gene Name: | protocadherin alpha 3 |
Gene Aliases: |
PCDH-ALPHA3 |
Location: |
Chr.5:140801198-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA3 | NM_018906.2 | NP_061729.1 | ||
| AF152311.1 | AAD43705.1 |
Target Gene Details
Entrez Gene ID: | 56144 |
Gene Name: | protocadherin alpha 4 |
Gene Aliases: |
CNR1, CNRN1, CRNR1, PCDH-ALPHA4 |
Location: |
Chr.5:140807074-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA4 | NM_018907.3 | NP_061730.1 | ||
| AB208925.1 | BAD92162.1 | |||
| AF152312.1 | AAD43706.1 |
Target Gene Details
Entrez Gene ID: | 56143 |
Gene Name: | protocadherin alpha 5 |
Gene Aliases: |
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5 |
Location: |
Chr.5:140821776-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA5 | NM_018908.2 | NP_061731.1 | ||
| AB208863.1 | BAD92100.1 | |||
| AF152313.1 | AAD43707.1 | |||
| BC033735.1 | AAH33735.1 |
Target Gene Details
Entrez Gene ID: | 56142 |
Gene Name: | protocadherin alpha 6 |
Gene Aliases: |
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6 |
Location: |
Chr.5:140827922-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA6 | NM_018909.3 | NP_061732.1 | ||
| NM_031849.2 | NP_114037.1 | |||
| AB209205.1 | BAD92442.1 | |||
| AF152314.1 | AAD43708.1 | |||
| AF169694.1 | AAF89691.1 | |||
| BC036674.2 | AAH36674.1 |
Target Gene Details
Entrez Gene ID: | 56141 |
Gene Name: | protocadherin alpha 7 |
Gene Aliases: |
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7 |
Location: |
Chr.5:140834384-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA7 | NM_018910.2 | NP_061733.1 | ||
| AF152315.1 | AAD43709.1 |
Target Gene Details
Entrez Gene ID: | 56140 |
Gene Name: | protocadherin alpha 8 |
Gene Aliases: |
PCDH-ALPHA8 |
Location: |
Chr.5:140841322-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA8 | NM_018911.2 | NP_061734.1 | ||
| AF152316.1 | AAD43710.1 |
Target Gene Details
Entrez Gene ID: | 9752 |
Gene Name: | protocadherin alpha 9 |
Gene Aliases: |
PCDH-ALPHA9 |
Location: |
Chr.5:140847772-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA9 | NM_031857.1 | NP_114063.1 | ||
| AF152317.1 | AAD43711.1 | |||
| AJ007610.1 | CAC22257.1 | |||
| AK314303.1 | ||||
| BC104802.1 | AAI04803.1 | |||
| BC104804.1 | AAI04805.1 |
Target Gene Details
Entrez Gene ID: | 56135 |
Gene Name: | protocadherin alpha subfamily C, 1 |
Gene Aliases: |
PCDH-ALPHA-C1 |
Location: |
Chr.5:140926717-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHAC1 | NM_018898.3 | NP_061721.2 | ||
| AF152303.1 | AAD43697.1 | |||
| AJ007608.1 | CAC22255.1 |
Target Gene Details
Entrez Gene ID: | 56134 |
Gene Name: | protocadherin alpha subfamily C, 2 |
Gene Aliases: |
PCDH-ALPHA-C2 |
Location: |
Chr.5:140966162-141012344 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHAC2 | NM_018899.5 | 1 | 1495 | NP_061722.1 |
| NM_031883.2 | 1 | 890 | NP_114089.1 | |
| AF152304.1 | 1 | 890 | AAD43698.1 | |
| AF152474.1 | 1 | 890 | AAD43735.1 | |
| AK123867.1 | 1 | 1422 | ||
| BC063288.1 | 1 | 1132 | AAH63288.1 | |
| BC104776.1 | 1 | 952 | AAI04777.1 | |
| BC105111.1 | 1 | 952 | AAI05112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv1031849 | Chr.5:140953984 - 141040639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map