Assay Details
Target Gene Details
Entrez Gene ID: | 871 |
Gene Name: | serpin family H member 1 |
Gene Aliases: |
AsTP3, CBP1, CBP2, HSP47, OI10, PIG14, PPROM, RA-A47, SERPINH2, gp46 |
Location: |
Chr.11:75562056-75572804 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINH1 | NM_001207014.1 | 1 | 139 | NP_001193943.1 |
| NM_001235.3 | 1 | 139 | NP_001226.2 | |
| AK290124.1 | 1 | 76 | ||
| CB123399.1 | 1 | 78 | ||
| DC398178.1 | 1 | 139 |
More Information
Set Membership: |
|

Genomic Map