Assay Details
Target Gene Details
Entrez Gene ID: | 6402 |
Gene Name: | selectin L |
Gene Aliases: |
CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1 |
Location: |
Chr.1:169690665-169711702 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SELL | NM_000655.4 | 9 | 1418 | NP_000646.2 |
| NR_029467.1 | 7 | 1182 | ||
| AJ246000.1 | 9 | 1289 | CAB55488.1 | |
| AK225713.1 | 9 | 1329 | ||
| AK298705.1 | 7 | 1177 | ||
| M25280.1 | 9 | 1329 | AAC63053.1 | |
| X16070.1 | 9 | 1342 | CAA34203.1 | |
| X16150.1 | 9 | 1305 | CAA34275.1 | |
| X17519.1 | 9 | 1302 | CAB43536.1 |
More Information
Set Membership: |
|

Genomic Map