Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map