Assay Details
Target Gene Details
Entrez Gene ID: | 124857 |
Gene Name: | WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 |
Gene Aliases: |
GASP-1, WFDC20B, WFIKKNRP, hGASP-1 |
Location: |
Chr.17:50834650-50842353 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFIKKN2 | NM_175575.5 | 2 | 2883 | NP_783165.1 |
| XM_017024172.1 | 2 | 2362 | XP_016879661.1 | |
| AK127743.1 | 2 | 2543 | ||
| AY358142.1 | 2 | 2718 | AAQ88509.1 | |
| BC140842.1 | ||||
| BC140844.1 |
More Information
Set Membership: |
|

Genomic Map