Assay Details
Target Gene Details
Entrez Gene ID: | 10758 |
Gene Name: | TRAF3 interacting protein 2 |
Gene Aliases: |
ACT1, C6orf2, C6orf4, C6orf5, C6orf6, CANDF8, CIKS, PSORS13 |
Location: |
Chr.6:111555378-111606874 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP2 | NM_001164281.2 | NP_001157753.1 | ||
| NM_147200.2 | NP_671733.2 | |||
| NM_147686.3 | NP_679211.2 | |||
| XM_011535386.1 | XP_011533688.1 | |||
| AF085921.1 | ||||
| AF136405.1 | AAF67445.1 | |||
| AF136406.1 | AAF67446.1 | |||
| AF136407.1 | AAF67447.1 | |||
| AF272151.1 | AAG15407.1 | |||
| AF274303.1 | AAG15367.1 | |||
| AK025351.1 | BAB15117.1 | |||
| AK310141.1 | ||||
| AK310922.1 | ||||
| AK314415.1 | ||||
| AL050289.1 | CAB43390.1 | |||
| BC002823.2 | AAH02823.1 | |||
| DA665549.1 | ||||
| DA836923.1 | ||||
| DC348710.1 |
Target Gene Details
Entrez Gene ID: | 643749 |
Gene Name: | TRAF3IP2 antisense RNA 1 |
Gene Aliases: |
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2 |
Location: |
Chr.6:111483472-111602295 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP2-AS1 | NR_034108.1 | |||
| NR_034109.1 |
More Information
Set Membership: |
|

Genomic Map