Assay Details
Target Gene Details
Entrez Gene ID: | 9429 |
Gene Name: | ATP binding cassette subfamily G member 2 (Junior blood group) |
Gene Aliases: |
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1 |
Location: |
Chr.4:88090264-88231417 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG2 | NM_001257386.1 | 16 | 3941 | NP_001244315.1 |
| NM_004827.2 | 16 | 4096 | NP_004818.2 | |
| XM_005263354.3 | 16 | 3939 | XP_005263411.1 | |
| XM_005263355.3 | 16 | 4046 | XP_005263412.1 | |
| XM_005263356.3 | 16 | 3816 | XP_005263413.1 | |
| XM_011532420.2 | 18 | 4210 | XP_011530722.1 | |
| XM_017008852.1 | 16 | 3933 | XP_016864341.1 | |
| XM_017008853.1 | 16 | 3736 | XP_016864342.1 | |
| BC092408.1 | 16 | 3941 | AAH92408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4413 | Chr.4:88083754 - 88129196 on Build GRCh38 | Deletion |
|
| esv2665331 | Chr.4:87937174 - 88153583 on Build GRCh38 | Deletion |
|
| esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map