Assay Details
Target Gene Details
Entrez Gene ID: | 3000 |
Gene Name: | guanylate cyclase 2D, retinal |
Gene Aliases: |
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC |
Location: |
Chr.17:8002670-8020340 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY2D | NM_000180.3 | 3 | 892 | NP_000171.1 |
| XM_011523816.1 | 2 | 818 | XP_011522118.1 | |
| M92432.1 | 3 | 891 | AAA60547.1 |
More Information
Set Membership: |
|

Genomic Map