Assay Details
Target Gene Details
Entrez Gene ID: | 6591 |
Gene Name: | snail family transcriptional repressor 2 |
Gene Aliases: |
SLUG, SLUGH1, SNAIL2, WS2D |
Location: |
Chr.8:48917677-48921440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAI2 | NM_003068.4 | 3 | 836 | NP_003059.1 |
| AK223368.1 | 3 | 825 | BAD97088.1 | |
| AK312661.1 | 3 | 825 | ||
| BC014890.1 | 3 | 813 | AAH14890.1 | |
| BC015895.1 | 3 | 793 | AAH15895.1 |
More Information
Set Membership: |
|

Genomic Map