Assay Details
Target Gene Details
Entrez Gene ID: | 56339 |
Gene Name: | methyltransferase like 3 |
Gene Aliases: |
IME4, M6A, MT-A70, Spo8 |
Location: |
Chr.14:21498131-21511375 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL3 | NM_019852.4 | 3 | 681 | NP_062826.2 |
| XM_006720206.3 | 3 | 661 | XP_006720269.1 | |
| XM_011536968.1 | 3 | 676 | XP_011535270.1 | |
| AK000222.1 | 3 | 604 | ||
| AK093509.1 | 1 | 463 | ||
| AK097873.1 | 2 | 1489 | ||
| AK098655.1 | 2 | 377 | ||
| AK300289.1 | 3 | 646 | ||
| AK304251.1 | 3 | 646 | ||
| AK304570.1 | ||||
| BC001650.2 | 3 | 615 | AAH01650.1 | |
| BC003031.1 | 3 | 615 | AAH03031.1 | |
| BC052244.1 | 3 | 565 | AAH52244.1 | |
| BX247964.1 | CAD62303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633718 | Chr.14:21495207 - 21527525 on Build GRCh38 | Gain |
|
| esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
|
| nsv85879 | Chr.14:21497546 - 21506056 on Build GRCh38 | Deletion |
|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map