Assay Details
Target Gene Details
Entrez Gene ID: | 3416 |
Gene Name: | insulin degrading enzyme |
Gene Aliases: |
INSULYSIN |
Location: |
Chr.10:92451684-92574484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDE | NM_001165946.1 | NP_001159418.1 | ||
| NM_001322793.1 | NP_001309722.1 | |||
| NM_001322794.1 | NP_001309723.1 | |||
| NM_001322795.1 | NP_001309724.1 | |||
| NM_001322796.1 | NP_001309725.1 | |||
| NM_001322797.1 | NP_001309726.1 | |||
| NM_004969.3 | NP_004960.2 | |||
| NR_136399.1 | ||||
| XM_017016187.1 | XP_016871676.1 | |||
| XM_017016188.1 | XP_016871677.1 | |||
| XM_017016189.1 | XP_016871678.1 | |||
| XM_017016190.1 | XP_016871679.1 | |||
| AB209204.1 | BAD92441.1 | |||
| AK093287.1 | ||||
| AK302578.1 | ||||
| AK312810.1 | ||||
| AK316407.1 | ||||
| BC096336.1 | AAH96336.1 | |||
| BC096337.1 | AAH96337.1 | |||
| BC096339.3 | AAH96339.1 | |||
| BX648462.1 | ||||
| M21188.1 | AAA52712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508600 | Chr.10:92355268 - 92472160 on Build GRCh38 | Deletion |
|
| dgv1349n54 | Chr.10:92452985 - 92530727 on Build GRCh38 | Loss |
|
| nsv1046258 | Chr.10:92107212 - 92706349 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map