Assay Details
Target Gene Details
Entrez Gene ID: | 1824 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
ARVD11, CDHF2, DG2, DGII/III, DSC3 |
Location: |
Chr.18:31065974-31102432 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSC2 | NM_004949.4 | 1 | 292 | NP_004940.1 |
| NM_024422.4 | 1 | 292 | NP_077740.1 | |
| AK290313.1 | ||||
| BC063291.1 | 1 | 274 | AAH63291.1 | |
| BX648425.1 | CAH56152.1 | |||
| DC382631.1 | 1 | 292 |
Target Gene Details
Entrez Gene ID: | 101927698 |
Gene Name: | DSC1/DSC2 antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.18:31101588-31162789 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAS | NR_110785.1 | |||
| DA156712.1 |
More Information
Set Membership: |
|

Genomic Map