Assay Details
Target Gene Details
Entrez Gene ID: | 79866 |
Gene Name: | bora, aurora kinase A activator |
Gene Aliases: |
C13orf34 |
Location: |
Chr.13:72727739-72756198 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORA | NM_001286746.1 | 10 | 1576 | NP_001273675.1 |
| NM_001286747.1 | 9 | 1371 | NP_001273676.1 | |
| NM_024808.3 | 10 | 1478 | NP_079084.3 | |
| XM_006719868.3 | 8 | 1335 | XP_006719931.2 | |
| AK026277.1 | 5 | 803 | BAB15426.1 | |
| AK291496.1 | 10 | 1304 | ||
| AK298608.1 | 9 | 1197 | ||
| BC007119.1 | 1 | 262 | AAH07119.1 | |
| BC025367.2 | 10 | 1289 | AAH25367.2 | |
| BC044213.1 | 9 | 1102 | AAH44213.1 | |
| BC056876.1 | 10 | 1301 | AAH56876.2 | |
| BX648605.1 | 10 | 1576 | ||
| EU834129.1 | 10 | 1384 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map