Assay Details
Target Gene Details
Entrez Gene ID: | 8328 |
Gene Name: | growth factor independent 1B transcriptional repressor |
Gene Aliases: |
BDPLT17, ZNF163B |
Location: |
Chr.9:132945545-132991697 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFI1B | NM_001135031.1 | NP_001128503.1 | ||
| NM_004188.6 | 5 | 732 | NP_004179.3 | |
| XM_011519068.2 | 3 | 623 | XP_011517370.1 | |
| XM_011519069.2 | 2 | 393 | XP_011517371.1 | |
| XM_011519070.2 | 3 | 623 | XP_011517372.1 | |
| XM_017015175.1 | 2 | 353 | XP_016870664.1 | |
| XM_017015176.1 | 3 | 623 | XP_016870665.1 | |
| AY428733.1 | AAR06639.1 | |||
| BC043371.1 | 5 | 732 | AAH43371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map