Assay Details
Target Gene Details
Entrez Gene ID: | 25915 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 3 |
Gene Aliases: |
2P1, C3orf60, E3-3 |
Location: |
Chr.3:49020475-49023495 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF3 | NM_199069.1 | 5 | 1308 | NP_951032.1 |
| NM_199070.1 | 5 | 871 | NP_951033.1 | |
| NM_199073.1 | 5 | 942 | NP_951047.1 | |
| NM_199074.1 | 5 | 823 | NP_951056.1 | |
| AA907148.1 | 1 | 80 | ||
| BM985326.1 | 1 | 87 |
More Information
Additional Information:
For this assay, SNP(s) [rs150913653] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map