Assay Details
Target Gene Details
Entrez Gene ID: | 387521 |
Gene Name: | transmembrane protein 189 |
Gene Aliases: |
KUA |
Location: |
Chr.20:50123737-50153798 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM189 | NM_001162505.1 | 6 | 1993 | NP_001155977.1 |
| NM_199129.2 | 6 | 2002 | NP_954580.1 | |
| NR_027889.1 | 7 | 2106 | ||
| AF155120.1 | 6 | 1929 | AAF08702.1 | |
| AK023028.1 | 6 | 1927 | BAB14364.1 |
Target Gene Details
Entrez Gene ID: | 387522 |
Gene Name: | TMEM189-UBE2V1 readthrough |
Gene Aliases: |
CROC-1B, CROC1B, KUA-UEV |
Location: |
Chr.20:50081124-50153798 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM189-UBE2V1 | NM_199203.2 | NP_954673.1 | ||
| U39361.1 | AAB72016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061982 | Chr.20:50111064 - 50483508 on Build GRCh38 | Loss |
|
| nsv834000 | Chr.20:50037107 - 50163145 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map