Assay Details
Target Gene Details
Entrez Gene ID: | 79048 |
Gene Name: | SECIS binding protein 2 |
Gene Aliases: |
SBP2 |
Location: |
Chr.9:89318473-89367117 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SECISBP2 | XM_011519000.2 | 18 | 3757 | XP_011517302.1 |
Target Gene Details
Entrez Gene ID: | 10507 |
Gene Name: | semaphorin 4D |
Gene Aliases: |
C9orf164, CD100, M-sema-G, SEMAJ, coll-4 |
Location: |
Chr.9:89360791-89498065 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA4D | NM_001142287.1 | 21 | 3362 | NP_001135759.1 |
| AJ420442.1 | 2 | 715 | ||
| AK090804.1 | 4 | 1358 | ||
| AK091601.1 | 4 | 3305 | ||
| AK097056.1 | 3 | 1609 | BAC04938.1 | |
| AK128342.1 | 2 | 2825 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| esv29434 | Chr.9:89348488 - 89728467 on Build GRCh38 | Gain+Loss |
|
| nsv1041327 | Chr.9:89298100 - 89438209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map