Assay Details
Target Gene Details
Entrez Gene ID: | 9382 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
CDG2G, LDLB |
Location: |
Chr.17:73193034-73208507 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG1 | NM_018714.2 | 1 | 99 | NP_061184.1 |
| AB037802.1 | 1 | 61 | BAA92619.1 | |
| AK098224.1 | 1 | 77 | ||
| BC021985.2 | 1 | 64 | AAH21985.1 | |
| BC047465.1 | 2 | 399 | AAH47465.1 | |
| BI667005.1 | 1 | 105 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2716191 | Chr.17:73192485 - 73193344 on Build GRCh38 | Deletion |
|
| esv2104946 | Chr.17:73192665 - 73193096 on Build GRCh38 | Deletion |
|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map