Assay Details
Target Gene Details
Entrez Gene ID: | 23094 |
Gene Name: | signal induced proliferation associated 1 like 3 |
Gene Aliases: |
CTRCT45, SPAL3, SPAR3 |
Location: |
Chr.19:37906823-38208372 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIPA1L3 | NM_015073.2 | NP_055888.1 | ||
| XM_017026518.1 | 1 | 422 | XP_016882007.1 | |
| BC036863.1 | AAH36863.1 | |||
| BC150620.1 | ||||
| BM193478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833820 | Chr.19:37699820 - 37923409 on Build GRCh38 | Loss |
|
| nsv1064401 | Chr.19:37897157 - 37937344 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map