Assay Details
Target Gene Details
Entrez Gene ID: | 4172 |
Gene Name: | minichromosome maintenance complex component 3 |
Gene Aliases: |
HCC5, P1-MCM3, P1.h, RLFB |
Location: |
Chr.6:52264014-52284881 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3 | NM_001270472.1 | 16 | 2549 | NP_001257401.1 |
| NM_002388.4 | 17 | 2662 | NP_002379.3 | |
| AI683709.1 | ||||
| AK222586.1 | 17 | 2500 | BAD96306.1 | |
| AK299566.1 | ||||
| AK300754.1 | 16 | 2375 | ||
| AK301704.1 | 17 | 2601 | ||
| AK312928.1 | ||||
| AY032603.1 | 17 | 2509 | AAK56392.1 | |
| BC001626.1 | 17 | 2507 | AAH01626.1 | |
| BC003509.1 | 17 | 2508 | AAH03509.2 | |
| D38073.1 | 17 | 2532 | BAA07267.1 | |
| KU178170.1 | ||||
| KU178171.1 | ||||
| X62153.2 | 17 | 2499 | CAA44078.2 |
More Information
Set Membership: |
|

Genomic Map