Assay Details
Target Gene Details
Entrez Gene ID: | 57697 |
Gene Name: | Fanconi anemia complementation group M |
Gene Aliases: |
FAAP250, KIAA1596 |
Location: |
Chr.14:45135939-45200890 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCM | NM_001308133.1 | 1 | 103 | NP_001295062.1 |
| NM_001308134.1 | 1 | 103 | NP_001295063.1 | |
| NM_020937.3 | 1 | 103 | NP_065988.1 | |
| XM_011537034.2 | 1 | 102 | XP_011535336.1 | |
| XM_011537035.2 | 1 | 102 | XP_011535337.1 | |
| XM_017021523.1 | 1 | 102 | XP_016877012.1 | |
| XM_017021525.1 | 1 | 102 | XP_016877014.1 | |
| XM_017021527.1 | 1 | 49 | XP_016877016.1 | |
| AK093422.1 | 1 | 102 | BAC04159.1 | |
| BC036056.1 | 1 | 104 | AAH36056.1 | |
| BC140776.1 | 1 | 88 | ||
| BC144511.1 | 1 | 88 | ||
| BX500546.1 | 1 | 110 | ||
| DQ140356.1 | 1 | 110 | AAZ53290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map