Assay Details
Target Gene Details
Entrez Gene ID: | 8522 |
Gene Name: | growth arrest specific 7 |
Gene Aliases: |
MLL/GAS7 |
Location: |
Chr.17:9910609-10198551 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS7 | NM_001130831.1 | 14 | 6469 | NP_001124303.1 |
| NM_003644.2 | 10 | 6218 | NP_003635.2 | |
| NM_201432.1 | 14 | 6626 | NP_958836.1 | |
| NM_201433.1 | 14 | 6656 | NP_958839.1 | |
| XM_005256831.3 | 15 | 6567 | XP_005256888.1 | |
| XM_005256832.3 | 14 | 6437 | XP_005256889.1 | |
| XM_005256833.3 | 13 | 6544 | XP_005256890.1 | |
| XM_005256834.3 | 13 | 6370 | XP_005256891.1 | |
| XM_005256835.3 | 13 | 6314 | XP_005256892.1 | |
| XM_011524044.2 | 14 | 6461 | XP_011522346.1 | |
| XM_017025239.1 | 14 | 6490 | XP_016880728.1 | |
| XM_017025240.1 | 13 | 6273 | XP_016880729.1 | |
| XM_017025241.1 | 13 | 6249 | XP_016880730.1 | |
| XM_017025242.1 | 13 | 6423 | XP_016880731.1 | |
| AB007854.1 | 14 | 6424 | ||
| AK225165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv38n68 | Chr.17:9897032 - 10093580 on Build GRCh38 | Loss |
|
| nsv516119 | Chr.17:9901176 - 9917378 on Build GRCh38 | Loss |
|
| nsv1065401 | Chr.17:9799828 - 9986825 on Build GRCh38 | Loss |
|
| nsv833361 | Chr.17:9785863 - 9979090 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map