Assay Details
Target Gene Details
Entrez Gene ID: | 60489 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3G |
Gene Aliases: |
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1 |
Location: |
Chr.22:39077005-39087743 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3G | NM_021822.3 | NP_068594.1 | ||
| XM_006724290.1 | XP_006724353.1 | |||
| XM_017028903.1 | XP_016884392.1 | |||
| XM_017028904.1 | XP_016884393.1 | |||
| AB266487.1 | ||||
| AF182420.1 | AAG14956.1 | |||
| AK022802.1 | ||||
| AK092614.1 | ||||
| AK093635.1 | ||||
| AK310279.1 | ||||
| AK315650.1 | ||||
| BC024268.1 | AAH24268.1 | |||
| CR456472.1 | CAG30358.1 | |||
| JF262036.1 | ||||
| JX915801.1 | ||||
| KC329531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834197 | Chr.22:39077294 - 39214999 on Build GRCh38 | Gain |
|
| nsv510807 | Chr.22:39075404 - 39147379 on Build GRCh38 | Deletion |
|
| nsv589032 | Chr.22:39077624 - 39086804 on Build GRCh38 | Loss |
|
| nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss |
|
| esv2724243 | Chr.22:39025861 - 39081892 on Build GRCh38 | Deletion |
|
| nsv1066354 | Chr.22:39070438 - 39130755 on Build GRCh38 | Gain |
|
| nsv955157 | Chr.22:39076996 - 39092195 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map