Assay Details
Target Gene Details
Entrez Gene ID: | 9158 |
Gene Name: | FGF1 intracellular binding protein |
Gene Aliases: |
FGFIBP, FIBP-1 |
Location: |
Chr.11:65883740-65888539 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIBP | NM_004214.4 | 10 | 1195 | NP_004205.2 |
| NM_198897.1 | 10 | 1216 | NP_942600.1 | |
| AF010187.2 | AAC97140.2 | |||
| AF171944.1 | AAD51694.1 | |||
| AF171945.1 | AAD51695.1 | |||
| AF171946.1 | AAD51696.1 | |||
| AF250391.1 | AAG01396.1 | |||
| AI925637.1 | 1 | 236 | ||
| AK223438.1 | BAD97158.1 | |||
| AK289562.1 | ||||
| BC014388.2 | AAH14388.1 | |||
| BC017448.1 | AAH17448.1 | |||
| CR456749.1 | CAG33030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv951019 | Chr.11:65874630 - 65899129 on Build GRCh38 | Deletion |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map