Assay Details
Target Gene Details
Entrez Gene ID: | 5652 |
Gene Name: | protease, serine 8 |
Gene Aliases: |
CAP1, PROSTASIN |
Location: |
Chr.16:31131433-31135762 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS8 | NM_002773.4 | 4 | 680 | NP_002764.1 |
| AK296656.1 | ||||
| AK301619.1 | 4 | 483 | ||
| AU142128.1 | ||||
| BC001462.2 | 4 | 591 | AAH01462.1 | |
| L41351.1 | 4 | 644 | AAC41759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map