Assay Details
Target Gene Details
Entrez Gene ID: | 56897 |
Gene Name: | Werner helicase interacting protein 1 |
Gene Aliases: |
WHIP, bA420G6.2 |
Location: |
Chr.6:2765341-2785745 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WRNIP1 | NM_020135.2 | 7 | 2080 | NP_064520.2 |
| NM_130395.2 | 7 | 2005 | NP_569079.1 | |
| AB056152.1 | 7 | 2081 | BAB60709.1 | |
| AB209723.1 | 7 | 1833 | BAD92960.1 | |
| AF218313.1 | 7 | 1467 | AAF80563.1 | |
| AK026179.1 | 6 | 985 | BAB15383.1 | |
| AK094823.1 | 6 | 1887 | ||
| AK223593.1 | 7 | 2014 | BAD97313.1 | |
| AK315047.1 | 7 | 2172 | ||
| AK315471.1 | 7 | 1974 | ||
| BC018923.2 | 7 | 1966 | AAH18923.1 | |
| U56249.1 | 3 | 325 |
More Information
Set Membership: |
|

Genomic Map