Assay Details
Target Gene Details
Entrez Gene ID: | 22937 |
Gene Name: | SREBF chaperone |
Gene Aliases: |
- |
Location: |
Chr.3:47413685-47477126 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCAP | NM_001320044.1 | 10 | 1378 | NP_001306973.1 |
| NM_012235.3 | 13 | 1863 | NP_036367.2 | |
| XM_005264967.1 | 13 | 1790 | XP_005265024.1 | |
| XM_005264968.1 | 10 | 1532 | XP_005265025.1 | |
| XM_005264971.1 | 7 | 943 | XP_005265028.1 | |
| XM_011533501.1 | 13 | 1890 | XP_011531803.1 | |
| XM_011533502.2 | 5 | 539 | XP_011531804.1 | |
| XM_017005918.1 | 13 | 1877 | XP_016861407.1 | |
| XM_017005919.1 | 13 | 1810 | XP_016861408.1 | |
| XM_017005920.1 | 13 | 1810 | XP_016861409.1 | |
| XM_017005921.1 | 7 | 943 | XP_016861410.1 | |
| AK027402.1 | 7 | 856 | BAB55088.1 | |
| AK075528.1 | BAC11673.1 | |||
| AK291484.1 | 8 | 1056 | ||
| AK300311.1 | 10 | 1268 | ||
| BC020987.1 | 5 | 584 | AAH20987.1 | |
| BC027207.2 | AAH27207.1 | |||
| D83782.2 | 13 | 1849 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
| nsv954480 | Chr.3:47404811 - 47421010 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map