Assay Details
Target Gene Details
Entrez Gene ID: | 51179 |
Gene Name: | hydroxyacid oxidase 2 |
Gene Aliases: |
GIG16, HAOX2 |
Location: |
Chr.1:119361913-119394130 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAO2 | NM_001005783.2 | NP_001005783.2 | ||
| NM_016527.3 | NP_057611.1 | |||
| XM_011541558.1 | XP_011539860.1 | |||
| XM_011541559.1 | XP_011539861.1 | |||
| XM_011541560.1 | XP_011539862.1 | |||
| XM_017001421.1 | XP_016856910.1 | |||
| AA919149.1 | ||||
| AF203975.1 | AAF14000.1 | |||
| AF231917.1 | AAF40200.1 | |||
| AK298289.1 | ||||
| AY513277.1 | AAT08030.1 | |||
| BC020863.1 | AAH20863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763647 | Chr.1:119386819 - 119612175 on Build GRCh38 | Gain+Loss |
|
| nsv2599 | Chr.1:119392400 - 119425125 on Build GRCh38 | Deletion |
|
| esv34089 | Chr.1:119209489 - 119530338 on Build GRCh38 | Loss |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
| nsv1004662 | Chr.1:119304824 - 119612175 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map