Assay Details
Target Gene Details
Entrez Gene ID: | 51603 |
Gene Name: | methyltransferase like 13 |
Gene Aliases: |
5630401D24Rik, CGI-01, KIAA0859, feat |
Location: |
Chr.1:171781504-171823962 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL13 | NM_001007239.1 | NP_001007240.1 | ||
| NM_014955.2 | 2 | 618 | NP_055770.1 | |
| NM_015935.4 | 2 | 943 | NP_057019.3 | |
| XM_017001455.1 | 1 | 437 | XP_016856944.1 | |
| XM_017001456.1 | 1 | 437 | XP_016856945.1 | |
| XM_017001457.1 | 1 | 437 | XP_016856946.1 | |
| XM_017001458.1 | 2 | 1060 | XP_016856947.1 | |
| XM_017001459.1 | 2 | 610 | XP_016856948.1 | |
| AB020666.2 | 2 | 698 | ||
| AB242174.1 | 2 | 868 | ||
| AB242175.1 | 2 | 856 | ||
| AF132936.1 | 2 | 868 | AAD27711.1 | |
| AK027552.1 | 2 | 862 | ||
| AK027621.1 | 2 | 557 | BAB55240.1 | |
| AK027886.1 | 1 | 414 | ||
| AK074552.1 | 1 | 347 | BAC11055.1 | |
| AK129561.1 | 1 | 292 | ||
| AK223493.1 | 2 | 904 | BAD97213.1 | |
| AK291740.1 | 2 | 890 | ||
| AK304471.1 | 3 | 724 | ||
| AL049669.1 | CAB41243.1 | |||
| BC029083.1 | 2 | 697 | AAH29083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522889 | Chr.1:171780525 - 171785931 on Build GRCh38 | Loss |
|
| nsv518222 | Chr.1:171783899 - 171794409 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map