Assay Details
Target Gene Details
Entrez Gene ID: | 7068 |
Gene Name: | thyroid hormone receptor beta |
Gene Aliases: |
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2 |
Location: |
Chr.3:24117153-24495281 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834639 | Chr.3:23995622 - 24192434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map