Assay Details
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3B | NM_001270449.1 | 6 | 998 | NP_001257378.1 |
| NM_001270450.1 | 6 | 653 | NP_001257379.1 | |
| NM_001270451.1 | 6 | 665 | NP_001257380.1 | |
| NM_130466.3 | 6 | 998 | NP_569733.2 | |
| NM_183415.2 | 6 | 653 | NP_904324.1 | |
| XM_005253987.2 | 6 | 878 | XP_005254044.1 | |
| XM_006719681.3 | 6 | 1211 | XP_006719744.1 | |
| XM_006719682.2 | 6 | 1211 | XP_006719745.1 | |
| XM_011538959.2 | 5 | 760 | XP_011537261.1 | |
| XM_011538961.1 | 6 | 1211 | XP_011537263.1 | |
| XM_017020195.1 | 6 | 866 | XP_016875684.1 | |
| XM_017020196.1 | 5 | 760 | XP_016875685.1 | |
| AF130072.1 | 4 | 390 | ||
| AF251046.2 | 6 | 968 | AAK28419.2 | |
| AK307385.1 | 5 | 532 | ||
| AL096740.1 | 6 | 967 | CAH56410.1 | |
| BC032301.1 | 6 | 631 | ||
| BC051266.1 | 6 | 969 | AAH51266.1 | |
| BC068221.1 | 6 | 635 | AAH68221.1 | |
| BC108705.1 | 6 | 629 | AAI08706.1 | |
| BC141880.1 | 6 | 876 | ||
| BC150637.1 | 6 | 629 | ||
| BX537403.1 | 6 | 643 | CAD97645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630719 | Chr.12:109485009 - 109700854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map