Assay Details
Target Gene Details
Entrez Gene ID: | 6279 |
Gene Name: | S100 calcium binding protein A8 |
Gene Aliases: |
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8 |
Location: |
Chr.1:153390032-153422583 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A8 | NM_001319196.1 | NP_001306125.1 | ||
| NM_001319197.1 | NP_001306126.1 | |||
| NM_001319198.1 | NP_001306127.1 | |||
| NM_001319201.1 | NP_001306130.1 | |||
| NM_002964.4 | NP_002955.2 | |||
| AA401538.1 | ||||
| AA903259.1 | ||||
| AI126134.1 | ||||
| AK291328.1 | ||||
| BC005928.2 | AAH05928.1 | |||
| BP209293.1 | ||||
| BP226540.1 | ||||
| BT007378.1 | AAP36042.1 | |||
| BU935536.1 | ||||
| CR407674.1 | CAG28602.1 | |||
| X06234.1 | CAA29580.1 | |||
| Y00278.1 | CAA68390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map