Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG2 | NM_001282211.1 | NP_001269140.1 | ||
| AK293514.1 |
Target Gene Details
Entrez Gene ID: | 122665 |
Gene Name: | ribonuclease A family member 8 |
Gene Aliases: |
RAE2 |
Location: |
Chr.14:21057822-21058455 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE8 | NM_138331.2 | 1 | 537 | NP_612204.1 |
| BC137537.1 | 1 | 537 | ||
| BC137538.1 | 1 | 537 | ||
| JQ353679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
| nsv1039369 | Chr.14:21030895 - 21062142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map