Assay Details
Target Gene Details
Entrez Gene ID: | 126133 |
Gene Name: | aldehyde dehydrogenase 16 family member A1 |
Gene Aliases: |
- |
Location: |
Chr.19:49453216-49471050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH16A1 | NM_001145396.1 | NP_001138868.1 | ||
| NM_153329.3 | NP_699160.2 | |||
| XM_011526441.1 | XP_011524743.1 | |||
| XM_011526442.1 | XP_011524744.1 | |||
| AK074136.1 | 15 | 3736 | BAB84962.1 | |
| AK297101.1 | ||||
| AK298587.1 | ||||
| AK301246.1 | ||||
| AY007096.1 | ||||
| BC014895.2 | AAH14895.2 | |||
| BC035641.1 | AAH35641.1 | |||
| BC042142.1 | AAH42142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| esv3644624 | Chr.19:49461676 - 49475440 on Build GRCh38 | Gain |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map