Assay Details
Target Gene Details
Entrez Gene ID: | 146223 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 4 |
Gene Aliases: |
CKLFSF4 |
Location: |
Chr.16:66598178-66696707 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM4 | NM_178818.2 | 5 | 3144 | NP_848933.1 |
| NM_181521.2 | 4 | 7847 | NP_852662.1 | |
| XM_011522882.1 | 4 | 7755 | XP_011521184.1 | |
| XM_017022954.1 | XP_016878443.1 | |||
| AF521889.1 | 5 | 3144 | AAN73845.1 | |
| AK000855.1 | 1 | 1023 | ||
| AL157468.1 | 1 | 1069 | ||
| AW207043.1 | 1 | 287 | ||
| CR933620.1 | 1 | 6756 |
More Information
Set Membership: |
|

Genomic Map