Assay Details
Target Gene Details
Entrez Gene ID: | 387597 |
Gene Name: | immunoglobulin like domain containing receptor 2 |
Gene Aliases: |
C1orf32, dJ782G3.1 |
Location: |
Chr.1:166913204-166975324 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILDR2 | NM_199351.2 | 9 | 1895 | NP_955383.1 |
| XM_006711314.3 | 4 | 2754 | XP_006711377.1 | |
| XM_017001252.1 | 9 | 2067 | XP_016856741.1 | |
| XM_017001253.1 | 9 | 2202 | XP_016856742.1 | |
| XM_017001254.1 | 8 | 2010 | XP_016856743.1 | |
| XM_017001255.1 | 8 | 1890 | XP_016856744.1 | |
| XM_017001256.1 | 6 | 1686 | XP_016856745.1 | |
| AF503509.1 | 9 | 1895 | AAQ07593.1 |
More Information
Set Membership: |
|

Genomic Map