Assay Details
Target Gene Details
Entrez Gene ID: | 57144 |
Gene Name: | p21 (RAC1) activated kinase 5 |
Gene Aliases: |
PAK7 |
Location: |
Chr.20:9537390-9839042 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK5 | NM_020341.3 | NP_065074.1 | ||
| NM_177990.2 | NP_817127.1 | |||
| XM_017027960.1 | XP_016883449.1 | |||
| XM_017027961.1 | XP_016883450.1 | |||
| XM_017027962.1 | XP_016883451.1 | |||
| XM_017027963.1 | XP_016883452.1 | |||
| XM_017027964.1 | XP_016883453.1 | |||
| XM_017027965.1 | XP_016883454.1 | |||
| AB033090.1 | ||||
| AB040812.1 | BAA94194.1 | |||
| AK095674.1 | ||||
| AK291401.1 | ||||
| AK307926.1 | ||||
| AK315820.1 | ||||
| AL133002.1 | CAB61350.1 | |||
| AX776013.1 | CAE11706.1 | |||
| BC024179.1 | AAH24179.1 |
More Information
Set Membership: |
|

Genomic Map