Assay Details
Target Gene Details
Entrez Gene ID: | 144100 |
Gene Name: | pleckstrin homology domain containing A7 |
Gene Aliases: |
- |
Location: |
Chr.11:16787153-17014423 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA7 | NM_175058.4 | 16 | 2264 | NP_778228.3 |
| XM_017017240.1 | 18 | 2472 | XP_016872729.1 | |
| XM_017017241.1 | 18 | 2472 | XP_016872730.1 | |
| XM_017017242.1 | 17 | 2352 | XP_016872731.1 | |
| XM_017017243.1 | 16 | 2271 | XP_016872732.1 | |
| AK296371.1 | 6 | 999 | ||
| AK301465.1 | 6 | 855 | ||
| BC033239.1 | 7 | 1141 | AAH33239.1 | |
| BC071599.1 | 16 | 2260 | AAH71599.1 |
More Information
Set Membership: |
|

Genomic Map